

Acetaldehyde oxime (mixture of syn and anti) ≥99%, Sigma-Aldrich®
Catalog # SIALA1002-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:59.07 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:203-479-6
UN:2332
:GHS02
:GHS07
CAS Number:107-29-9
Formula:C₂H₅NO
MDL Number:MFCD00002124