Order Entry
Poland
ContactUsLinkComponent
Phenylphosphine 10% in hexane, Sigma-Aldrich®
Phenylphosphine 10% in hexane, Sigma-Aldrich®
Catalog # SIAL674389-50ML
Supplier:  Merck
CAS Number:  
Phenylphosphine 10% in hexane, Sigma-Aldrich®
Catalog # SIAL674389-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :110.1 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:211-325-4
CAS Number:638-21-1
Formula:C₆H₇P
MDL Number:MFCD00002094