Order Entry
Poland
Orders LinkContactUsLinkComponent
Pterostilbene

Pterostilbene

Supplier:   Cayman Chemical
Pterostilbene
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
  • CAS Number:537-42-8
  • Formula:C₁₆H₁₆O₃
  • MDL Number:MFCD00238710
  • Molecular Weight:256.2964 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 419 zł
    419 zł
    1 * 50 mg
       419 zł

  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 791 zł
    791 zł
    1 * 100 mg
       791 zł

  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 1 670 zł
    1 670 zł
    1 * 250 mg
       1 670 zł

  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 3 130 zł
    3 130 zł
    1 * 500 mg
       3 130 zł