4-Hydroxy-3-methoxybenzonitrile, Sigma-Aldrich®
Catalog # SIAL162604-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:149.15 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:224-589-0
UN:3439
CAS Number:4421-08-3
Formula:C₈H₇NO₂
MDL Number:MFCD00001820