Order Entry
Poland
ContactUsLinkComponent
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV), Sigma-Aldrich®
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV), Sigma-Aldrich®
Catalog # SIAL478865-5G
Supplier:  Merck
CAS Number:  
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV), Sigma-Aldrich®
Catalog # SIAL478865-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :824.29 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:18865-74-2
Formula:C₄₄H₇₆O₈Zr
MDL Number:MFCD00145380