Order Entry
Poland
Orders LinkContactUsLinkComponent
avantor_acids95500.jpg
Avantor high-purity acids
The success of the application, reliability of results, and proper testing of trace metals all depend on the correct quality and grade of acid. Avantor J.T.Baker® and Macron Fine Chemicals™ brand high-purity acids provide a range of choices that are suitable for different levels of detection – even down to low, parts-per-trillion (ppt) levels.
Avantor high-purity acids | Avantor
Product Selection
avantor_Ultrex-II_90.jpg
Recommended for use in your most demanding trace element analyses; Analyzed for up to 65 trace elements in the low ppt range.
avantor_BIA_90.jpg
Recommended for ICP-OES/AES and GFAA applications, and other applications requiring ppb trace metal testing.
avantor_ACS_90.jpg
Recommended for qualitative AAS applications, as well as general wet chemistry.
avantor_MacronAR_90.jpg
Macron Fine Chemicals™ acids generate highly reliable and consistent parts-per-billion (ppb) trace metal analysis results.