anti-gas tablets using simethicone
Simethicone
NuSil® simethicone fluids and granular solids are used in a broad range of healthcare and pharmaceutical applications for their anti-gas and anti-foaming properties.
For a broad range of antacid and anti-gas treatments, the NuSil simethicone portfolio offers products designed for efficient use in both aqueous and non-aqueous solutions. NuSil simethicone is supplied in both liquid and granular solid options:
  • Simethicone fluids: Our formulations provide versatility in pharmaceutical applications with properties such as varying polymer viscosities, low volatility and resistance to silica separation. Made with superior bubble-coalescing activity, they are highly effective for the elimination and control of foam in both aqueous and non-aqueous solutions.
  • Simethicone granular solid: The fastest granular simethicone defoamer on the market, it has a proven history of use in chewable and soft-chew antacid tablets. It allows for easy blending with other tablet ingredients and is designed to reduce the cost of cleanup and additional equipment required for liquid addition of simethicone-active ingredients.
Customization mastered
No matter what kind of silicone product you are formulating, NuSil has the expertise, processes and proprietary technology to customize our silicone polymers and catalysts based on your unique specifications. We leverage our insight into the unique silicone chemistry and material properties to supply both off-the-shelf and custom formulations tailored to your process and end application performance requirements.
Learn more about how we can help you get the products you need.
Contact us
Interior NuSil silicone production technology
Product finder
General healthcare
NuSil simethicones provide a high-quality, versatile material for use in a broad range of anti-gas and antacid tablets. It is also an excellent defoaming agent used in biopharma processing.
Related content
Browse additional resources related to simethicones.
Customer support
Contact us
Ready to move your next innovation forward? NuSil can help.
How can we help you?
Whether you’re producing antacid and anti-gas products, or you need a proven defoaming agent, NuSil experts will help you find the right simethicone for your application.
Avantor®, a Fortune 500 company, is a leading global provider of mission-critical products and services to customers in the biopharma, healthcare, education & government, and advanced technologies & applied materials industries. Our portfolio is used in virtually every stage of the most important research, development and production activities in the industries we serve. One of our greatest strengths comes from having a global infrastructure that is strategically located to support the needs of our customers. Our global footprint enables us to serve more than 225,000 customer locations and gives us extensive access to research laboratories and scientists in more than 180 countries. We set science in motion to create a better world.