Class VI Line
SIL-5950
CLASS VI SUPPORTED LIQUID SILICONE RUBBER
Description
  • Two-part, translucent silicone system designed for use with injection molding equipment
  • Rapid cure with no post-cure required
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For the injection molding of parts requiring a material with a high durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
  • Suitable for over-molding applications
  • Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
NuSil Technology's SIL-5950 shall not be considered for use in human implantation for a period of greater than 29 days.
Regulatory Support
NuSil Technology’s Class VI (C6) products meet the requirements defined in ISO-10993 for Surface Devices with “limited” (<24 hours) or “prolonged” (1 to 30 days) contact duration.
After post-cure, material meets or exceeds the extraction requirements for FDA regulation 21 CFR 177.2600 “Rubber Articles Intended for Repeated Use (Food Contact) NuSil Technology’s SIL-5950 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 50 Type A
Tensile 1,450 psi (10 MPa)
Elongation 550 %
Tear 190 ppi (33.51 kN/m)
Appearance Translucent
Cure 5 minutes / 165 °C
Cure Rate T10 0.12 minutes
Cure System Platinum
Extrusion Rate 70 g/minute
Mix Ratio 1:1
Specific Gravity 1.14
Stress at Strain 400 psi / 200 % (2.76 MPa / 200 %)
Work Time
Comment Designed for High Volume Injection Molding
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.