SCV-2596
ULTRA LOW OUTGASSING ELECTRICALLY CONDUCTIVE RTV SILICONE
Description
  • Two-part, black electrically conductive RTV silicone
  • 10:1 Mix Ratio (Part A:B)
Applications
  • For applications requiring Ultra Low OutgassingTM and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
  • Use for RFI and EMI shielding in electronic and space applications
  • Use to adhere covers onto housings or for any application where grooves and other configurations require a flowable to limited flow material
  • For applications requiring a broader operating temperature range
Exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤0.1% and CVCM of ≤0.010%
Product Details
Property Average Result
Volume Resistivity 2.5 ohmcm
Cure System Platinum
Durometer 75 Type A
Tensile 475 psi (3.28 MPa)
Appearance Black
Cure 30 minutes / 150 °C
Elongation 90 %
Lap Shear 250 psi (1.72 MPa)
Lap Shear 250 psi (1.72 MPa)
Mix Ratio 10:1
Special Feature
Specific Gravity 1.19
Work Time 2 hours
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.