Description
- Two-part, red, low specific gravity, RTV silicone
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring Ultra Low OutgassingTM and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use a bonding, sealing, or potting material in electronic and space applications
- Provides radiation resistance, low thermal conductivity, oxidation stability, thermal stability and good ablative characteristics
- Especially useful to bond solar cells to solar array panels
- For applications requiring a broader operating temperature range
Exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a maximum TML of 0.1% and CVCM of less than 0.010%
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 325,000 cP (325,000 mPa*s) |
Lap Shear | 175 psi (1.21 MPa) |
Durometer | 45 Type A |
Appearance | Red |
Cure | 30 minutes / 150 °C |
Elongation | 150 % |
Mix Ratio | 1:1 |
Special Feature | |
Specific Gravity | 0.74 |
Tensile | 225 psi (1.55 MPa) |
Work Time | 4 hours |
Comment | Fast Cure, Low Density, Primed Lap Shear 175psi |
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.