Description
- Two-part, translucent silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- Convenient 1:1 mix ratio (Part A:Part B)
Applications
- For electronic and space applications requiring Ultra Low OutgassingTM and minimal volatile condensables under extreme operating conditions
- To provide protection of electronic components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For applications requiring a broader operating temperature range
Exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤0.1% and CVCM of ≤0.010%
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 77,000 cP (77,000 mPa*s) |
Lap Shear | 475 psi (3.28 MPa) |
Durometer | 35 Type A |
Appearance | Translucent |
Cure | 15 minutes / 150 °C |
Elongation | 300 % |
Mix Ratio | 1:1 |
Specific Gravity | 1.11 |
Tack Free Time | 7 hours |
Tear | 40 ppi (7.05 kN/m) |
Tensile | 700 psi (4.83 MPa) |
Work Time | 1 hours |
Comment | Ultra low Outgassing. Recommended for broad temperature applications. 475 psi (3.3 MPa), primed (CF1-135) lap shear. |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.