R7-3521-11

FLUOROSILICONE ELASTOMER

Description

  • Two-part, dark gray, 50 mole % fluorosilicone elastomer
  • 1:1 Mix Ratio (Part A:B)
  • Cures at room temperature or rapidly with heat

Applications

  • For applications requiring solvent resistance
  • For molding parts such as O-rings, stoppers, and seals

Product Details

Property Average Result
Cure System Platinum
Durometer 30 Type A
Appearance Gray
Attribute
Cure 48 hours / RTV
Elongation 260 %
Mix Ratio 1:1
Operating Temperature
Specific Gravity 1.27
Tear 35 ppi (6.17 kN/m)
Tensile 500 psi (3.45 MPa)
Work Time 1 hours
Comment Solvent Resistace

Making the right choice for fuel- and solvent-resistant silicones

Learn how high-purity fluorosilicones resist degradation and maintain performance in aviation and defense applications.

Read more

Save to favorites

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.