Description
- One-part RTV silicone elastomer dispersed in tert butyl acetate
- Based on a fluorosilicone copolymer
Applications
- For coating, sealing and bonding applications requiring solvent and/or fuel resistance
- Designed for spraying, dip casting, and RTV curing of thin, elastomeric films
- For use as a coating to minimize ice adhesion
- Bonds aggressively to most surfaces
- Slight addition of heat will accelerate cure
Product Details
| Property | Average Result |
|---|---|
| Viscosity | 1,625 cP (1,625 mPa*s) |
| Non Volatile Content | 60 % |
| Appearance | Translucent |
| Attribute | |
| Cure | 72 hours / RTV |
| Cure System | Acetoxy |
| Durometer | 25 Type A |
| Elongation | 400 % |
| Mix Ratio | 1 PART |
| Specific Gravity | 1.29 |
| Tear | 35 ppi (6.17 kN/m) |
| Tensile | 425 psi (2.93 MPa) |
| Comment | Dispersion Coating, 60% Solids |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.