Description
- Two-part, electrically conductive RTV Silicone
- Designed for enhanced performance in extreme low and high temperatures
- 100:0.5 Mix Ratio (Base:Curing Agent)
Applications
- For RFI and EMI shielding for electrical and aerospace applications
- Excellent for formed-in-place conductive gaskets and grounding connections
- Use to adhere covers on housings or where grooves or other configurations require a non-flowable to limited flow material
Product Details
| Property | Average Result |
|---|---|
| Cure System | Alkoxy |
| Extrusion Rate | 160 g/minute |
| Lap Shear | 200 psi (1.38 MPa) |
| Durometer | 80 Type A |
| Appearance | Gray |
| Cure | 7 days / RTV |
| Elongation | 90 % |
| Flow Rate | 1.2 in (30.48 mm) |
| Mix Ratio | 100:0.5 |
| Specific Gravity | 3.41 |
| Tear | 50 ppi (8.82 kN/m) |
| Tensile | 250 psi (1.72 MPa) |
| Volume Resistivity | 0 ohmcm |
| Work Time | 3 hours |
| Comment | 200 psi lap shear primed w/SP-121. Conductive at elevated temperatures. |
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.