R-2370
RTV SILICONE FOAM
Description
  • Two-part, RTV silicone foam
  • Yields a low density silicone rubber foam when catalyzed
  • 100:6 Mix Ratio (Base: Catalyst)
Applications
  • Use in shock and vibration dampening situations requiring a lightweight, flexible foam with excellent thermal and electrical properties
Product Details
Property Average Result
Viscosity 4,700 cP (4,700 MPa*s)
Cure System Tin
Foam Density 10 lbs/ft3 (0.16 g/cm3)
Application Life 28 minutes
Appearance Tan
Cure 10 minutes / RTV
Mix Ratio 100:6
Non Volatile Content 100 %
Comment Low Density / Soft, 10 lb/ft^3 (0.16 g/cm^3)
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.