Description
- Two-part, translucent, pourable silicone system
- 10:1 Mix Ratio (Part A:B)
- Cures at room temperature or rapidly with heat
- High tear strength and good physical properties
Applications
- To provide protection of electronic components and assemblies against shock, vibration, moisture, ozone, dust, chemicals and other environmental hazards
- Ideal for molding or use in O-rings, potting connectors, cable harness breakouts, molded high voltage terminals, seals and gaskets due to its high physical strength
- For applications requiring an operating temperature range of -65 to 240°C (-85 to 465°F)
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 83,000 cP (83,000 mPa*s) |
Lap Shear | 475 psi (3.28 MPa) |
Durometer | 30 Type A |
Appearance | Translucent |
Cure | 15 minutes / 150 °C |
Dielectric Strength | 640 volts/mil (24.96 kV/mm) |
Elongation | 450 % |
Mix Ratio | 10:1 |
Operating Temperature | -65 °C / 240 °C |
Specific Gravity | 1.12 |
Tear | 100 ppi (17.64 kN/m) |
Tensile | 1,050 psi (7.24 MPa) |
Work Time | 2.5 hours |
Comment | Legacy general all purpose adhesive. 475 psi (3.3 MPa), primed (CF1-135) lap shear. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.