R-2165
THERMALLY CONDUCTIVE SILICONE POTTING ELASTOMER
Description
  • Two part, Gray, 1:1 Mix ratio (A:B)
  • Pourable and self-leveling
  • Cures at room temperature
  • REACh and RoHS compliant
Applications
  • Enhanced thermal conductivity for removal of heat and improved reliability
  • Low viscosity allows the potting of complex geometries without the entrapment of air
  • Quick set-up and flexible cure schedule
  • Applications include: potting for sensors, relays, and other electronic components requiring protection from vibration and generally harsh environmental conditions
Product Details
Property Average Result
Thermal Conductivity 0.5 W/(mK)
Viscosity 4,000 cP (4,000 mPa*s)
Cure System Platinum
Tensile 500 psi (3.45 MPa)
Appearance Gray
Cte
Cure 10 minutes / 150 °C
Dielectric Strength 700 volts/mil (27.3 kV/mm)
Durometer 60 Type A
Elongation 100 %
Mix Ratio 1:1
Volume Resistivity 1,000,000,000,000,000 ohmcm
Work Time 10 minutes
Comment 0.6 W/mK, versatile, moderate thermally conductive silicone. Will RTV.
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.