Description
- Two-part, gray silicone adhesive system
- Cures at room temperature or rapidly by applying heat in an oven or by a heat gun or lamp
- Does not require atmospheric moisture to cure
- No curing byproducts such as acetic acid or methyl alcohol
- Product available in easy-to-use, airless side-by-side kits that eliminate mixing and deairing difficulties
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Adhesive for bonding and sealing silicones to each other and other substrates such as metals and plastics
- For rapid production or prototyping due to a rapid cure
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Extrusion Rate | 180 g/minute |
| Lap Shear | 145 psi (1 MPa) |
| Durometer | 45 Type A |
| Appearance | Dark Gray |
| Cure | 2 hours / 65 °C |
| Dielectric Strength | 825 volts/mil (32.18 kV/mm) |
| Elongation | 400 % |
| Mix Ratio | 1:1 |
| Specific Gravity | 1.17 |
| Stress at Strain | 525 psi / 200 % (3.62 MPa / 200 %) |
| Tear | 150 ppi (26.46 kN/m) |
| Tensile | 1,050 psi (7.24 MPa) |
| Work Time | 15 minutes |
| Comment | Primerless Adhesive;Gray, tough, fast cure adhesive. Will RTV. 145 psi (1.0 MPa), unprimed and 600 psi (4.1 MPa) primed (CF1-135) lap shear. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.