R-2100-1
SILICONE INK (WHITE)
Description
  • Strong adhesion to other silicones
  • Dispersed in Xylene
  • Accelerated cure with introduction of heat
  • Cures via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • Ideal for use as a marking ink for silicone parts and other components on which the coating must maintain long-term stability
  • Use in applications requiring a rapid cure schedule
  • Provides high opacity for single print applications
Product Details
Property Average Result
Cure System Platinum
Viscosity 560 cP (560 mPa*s)
Non Volatile Content 65 %
Mix Ratio 1:1
Appearance White
Tack Free Time 24 hours
Comment Available in white (R-2100-1), black (R-2100-2), red (R-2100-3), and green (R-2100-6). Dispersed in xylene.
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.