Description
- One-part, black, platinum catalyzed silicone Glob-Top
- Thixotropic consistency
Applications
- Provides protection for electronic assemblies
- Ensures microchip reliability by providing vital protection
- For applications that require precise delivery configurations
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 400,000 cP (400,000 mPa*s) |
Extrusion Rate | 120 g/minute |
Durometer | 19 Type A |
Appearance | Black |
Cure | 15 minutes / 200 °C |
Dielectric Strength | 400 volts/mil (15.6 kV/mm) |
Elongation | 750 % |
Mix Ratio | 1 PART |
Specific Gravity | 1.18 |
Stress at Strain | 50 psi / 100 % (0.34 MPa / 100 %) |
Tear | 110 ppi (19.4 kN/m) |
Tensile | 825 psi (5.69 MPa) |
Comment | Thixotropic, dispensable, and 0.5mm maximum slump. Shipped and stored frozen. |
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.