R-1130
NON-CORROSIVE RTV SILICONE ADHESIVE/SEALANT
Description
  • One-part, translucent, non-slumping RTV silicone adhesive
  • Based on a dimethyl silicone polymer
  • Non-corrosive cure
  • Shelf-life stability
  • Rapid curing
  • Excellent electrical insulation properties
Applications
  • As a sealing and caulking, adhesive or potting material for environmental protection of electronic assemblies and components where a non-corrosive cure is essential
  • As a coating for semi-conductors and integrated circuits
  • As an adhesive/sealant for electrical wires, terminals, mounting resistors and connectors
  • Ideal for overhead or side wall applications due to its non-slumping characteristics
Product Details
Property Average Result
Cure System Oxime
Extrusion Rate 70 g/minute
Lap Shear 485 psi (3.34 MPa)
Durometer 35 Type A
Appearance Translucent
Cure 7 days / RTV
Elongation 325 %
Flow Rate 0.5 in (12.7 mm)
Mix Ratio 1 PART
Non Volatile Content 100 %
Specific Gravity 1.1
Tack Free Time 25 minutes
Tear 40 ppi (7.05 kN/m)
Tensile 850 psi (5.86 MPa)
Comment Non-slump. Adhesive for a variety of substrates on or near electronic components. 485 psi (3.6 MPa), primed (SP-120) lap shear.
Specialty Silicones for Displays
The NuSil® line of optoelectronic silicones enable device manufacturers from smart phones to stadium screens and everything in between to create next-generation displays that are sharper, brighter and more rugged.