Description
- One-part RTV silicone elastomer dispersed in xylene
Applications
- Designed for spraying, dip casting, and room temperature curing of thin, elastomeric films
Product Details
Property | Average Result |
---|---|
Viscosity | 700 cP (700 mPa*s) |
Non Volatile Content | 29 % |
Appearance | Translucent |
Cure | 5 days / RTV |
Cure System | Acetoxy |
Durometer | 25 Type A |
Elongation | 950 % |
Refractive Index | 1.41 |
Specific Gravity | 1.09 |
Stress at Strain | 160 psi / 300 % (1.1 Mpa / 300 %) |
Tear | 125 ppi (22.05 kN/m) |
Tensile | 1,425 psi (9.83 MPa) |
Comment | Great adhesion to a variety of substrates. Not recommended where corrosion is of concern. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.