Description
- High-Strength, flowing, one-part RTV silicone rubber dispersed in VM&P Naphtha
- Cures to a transparent film within 24 hours
- Excellent release characteristics
- Comes in pigmented versions upon request
Applications
- As a protective coating to prevent corrosion and erosion of various surfaces in missiles, rockets, aircraft and launch equipment
- For making conformal bags used in the manufacturing of composite parts
- As a conformal coating for electronic applications
Product Details
Property | Average Result |
---|---|
Viscosity | 6,150 cP (6,150 mPa*s) |
Non Volatile Content | 32 % |
Appearance | Translucent |
Tack Free Time | 1.5 hours |
Cure | 7 days / RTV |
Cure System | Oxime |
Durometer | 45 Type A |
Elongation | 650 % |
Specific Gravity | 1.1 |
Tear | 95 ppi (16.75 kN/m) |
Tensile | 1,150 psi (7.93 MPa) |
Comment | High strength coating recommend for coating PCBs and other electronic assemblies. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.