Description
- One-part, non-curing, pressure sensitive adhesive
- 1.43 Refractive Index
Applications
- For temporarily adhering to various substrates including: metals, glass, silicone, and organic rubbers
Product Details
Property | Average Result |
---|---|
Viscosity | 2,100 cP (2,100 mPa*s) |
Cure | 30 minutes / 70 °C |
Non Volatile Content | 50 % |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.