Description
- A 30 mole % diphenylsiloxane endblocked with reactive vinyl groups
Applications
- For curing with crosslinkers and catalysts to form adhesives, sealant, rubbers and gels due to their reactive nature
Product Details
Property | Average Result |
---|---|
Refractive Index | 1.52 |
Viscosity | 575 cSt (619.14 mPa*s) |
Mmol | 0.8 Vi/g |
Appearance | Translucent |
Specific Gravity | 1.08 |
Volatile Content | 0.2 % |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.