PLY-7602
DIPHENYLMETHYLSILANOL
Description
- A functional diphenylmethylsilanol
Applications
- As processing aid or plasticizer for elastomeric systems
Product Details
Property | Average Result |
---|---|
Appearance | Clear |
Cure System | Non-Curing |
Mix Ratio | 1 PART |
Non Volatile Content | 52% |
Rheology | Flowable / Self Leveling |
Special Feature | Phenyl processing aid |
Comment | A processing aid or plasticizer for elastomeric systems |
Contact NuSilâ„¢ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.