Description
- A clear, dimethyl polysiloxane endblocked with vinyl dimethyl groups, with 7.5 mole % vinyl groups along the chain backbone for increased crosslinking ability
- Other viscosities and percentages of vinyl are available on a special order basis
Applications
- For curing with crosslinkers and catalysts to form adhesives, sealant, rubbers and gels due to its reactive nature
Product Details
Property | Average Result |
---|---|
Refractive Index | 1.41 |
Viscosity | 1,000 cSt (1,006.97 mPa*s) |
Mmol | 1.1 Vi/g |
Appearance | Translucent |
Specific Gravity | 1.01 |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.