MED4-4900
AMMONIUM BICARBONATE HEALTHCARE SILICONE FOAM MASTERBATCH
Description
- A single component masterbatch, with the consistency of a low viscosity paste
- A masterbatch consisting of ammonium bicarbonate dispersed in a vinyldimethyl-terminated polydimethylsiloxane polymer
Applications
- For use in producing silicone foams in heat curable silicone elastomer systems
Product Details
| Property | Average Result |
|---|---|
| Appearance | White |
| Mix Ratio | 1 PART |
| Comment | Ammonium Bicarbonate LSR Foam Masterbatch |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
Save to favorites
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.