Description
- A white, single component masterbatch, with the consistency of a clay like material
- Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
Applications
- For easy and more precise pigmentation of high consistency silicone elastomers, suitable for molding, calendering or extrusion
- Reduces production time and eliminates use of powders that may contaminate clean room environments
NuSil Technology’s MED3-4502 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Appearance | White |
| Mix Ratio | 1 PART |
| Comment | HCR Masterbatch, 75% TiO2 |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.
Save to favorites
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.