Premium Care Line
MED10-5440
LIQUID INJECTION MOLDING FLUOROSILICONE LIQUID SILICONE RUBBER
Description
- Two-part, translucent, fluorosilicone elastomer used with liquid injection molding equipment100 mol % fluorosiliconeCures with heat via addition-cure chemistry1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a medium durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
- Suitable for over-molding applications
NuSil MED10-5440 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Durometer | 30 Type A |
Tensile | 1,250 psi (8.62 MPa) |
Elongation | 380 % |
Tear | 60 ppi (10.58 kN/m) |
Appearance | Translucent |
Cure | 30 minutes / 150 °C |
Cure System | Platinum |
Extrusion Rate | 40 g/minute |
Mix Ratio | 1:1 |
Specific Gravity | 1.38 |
Work Time | |
Comment | 100M% Fluorosilicone Injection Molding Elastomer |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.