Implant Line
MED1-4502-2
COLOR MASTERBATCH FOR HIGH CONSISTENCY SILICONE ELASTOMER
Description
- A black, single component masterbatch, with the consistency of a clay like material
- Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
Applications
- For easy and more precise pigmentation of high consistency silicone materials, suitable for molding, calendering or extrusion
- Reduces production time and eliminates use of powders that may contaminate clean room environments
- Strict quality controls ensure color consistency
NuSil Technology’s MED1-4502-2 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Appearance | Black |
Mix Ratio | 1 PART |
Comment | Masterbatch to add color to HCR platinum cure systems |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.