Description
- Two-part silicone elastomer dispersed in xylene
- Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
- To provide a heat-cured, low-friction coating on cured silicone substrates
- For tubing (ID/OD). Balloons, valves, stoppers, o-rings, and for moving or sliding parts
NuSil™ MED-6670 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Non Volatile Content | 25 % |
Appearance | Translucent |
Cure | 5 minutes / RTV |
Mix Ratio | 1:1 |
Special Feature | |
Specific Gravity | 0.96 |
Work Time | 24 hours |
Comment | Low Coefficient of Friction Coating |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.