Description
- Two-part silicone elastomer dispersed in xylene
- Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
- To provide a heat-cured, low-friction coating on cured silicone substrates
- For tubing (ID/OD). Balloons, valves, stoppers, o-rings, and for moving or sliding parts
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Non Volatile Content | 25 % |
| Appearance | Translucent |
| Cure | 5 minutes / RTV |
| Mix Ratio | 1:1 |
| Special Feature | |
| Specific Gravity | 0.96 |
| Work Time | 24 hours |
| Comment | Low Coefficient of Friction Coating |
Contact NuSil® to Order Now
For over 20 years, the NuSil® brand has offered a wide range of silicone lubricants for use in medical devices. Our team is ready to collaborate with you to provide medical grade lubricious silicone solutions to satisfy your unique force reduction and material requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.