Description
- Two-part silicone elastomer dispersed in xylene
- Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
- To provide a heat-cured, low-friction coating on cured silicone substrates
- For tubing (ID/OD). Balloons, valves, stoppers, o-rings, and for moving or sliding parts
NuSil™ MED-6670 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Non Volatile Content | 25 % |
| Appearance | Translucent |
| Cure | 5 minutes / RTV |
| Mix Ratio | 1:1 |
| Special Feature | |
| Specific Gravity | 0.96 |
| Work Time | 24 hours |
| Comment | Low Coefficient of Friction Coating |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.