Description
- One-part, 100 mol % fluorosilicone, RTV elastomer dispersed in Tert-Butyl Acetate
Applications
- For dip casting of thin elastomeric films
- For sealing and bonding applications
- For applications requiring resistance (low swell/low permeation) to many solvents and oils including dimethyl silicone oils
NuSil Technology’s MED-6655 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure System | Acetoxy |
Viscosity | 700 cP (700 mPa*s) |
Non Volatile Content | 60 % |
Durometer | 35 Type A |
Appearance | White |
Cure | 3 days / RTV |
Cure Type | RTV |
Elongation | 425 % |
Mix Ratio | 1 PART |
Rheology | Flowable / Self Leveling |
Tack Free Time | 80 minutes |
Tear | 45 ppi (7.94 kN/m) |
Tensile | 775 psi (5.34 MPa) |
Comment | 100M% Fluorosilicone |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.