Description
- One-part, 100 mol % fluorosilicone, RTV elastomer dispersed in Tert-Butyl Acetate
Applications
- For dip casting of thin elastomeric films
- For sealing and bonding applications
- For applications requiring resistance (low swell/low permeation) to many solvents and oils including dimethyl silicone oils
NuSil Technology’s MED-6655 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Cure System | Acetoxy |
| Viscosity | 700 cP (700 mPa*s) |
| Non Volatile Content | 60 % |
| Durometer | 35 Type A |
| Appearance | White |
| Cure | 3 days / RTV |
| Cure Type | RTV |
| Elongation | 425 % |
| Mix Ratio | 1 PART |
| Rheology | Flowable / Self Leveling |
| Tack Free Time | 80 minutes |
| Tear | 45 ppi (7.94 kN/m) |
| Tensile | 775 psi (5.34 MPa) |
| Comment | 100M% Fluorosilicone |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.