Description
- Two-part medium viscosity silicone elastomer dispersed xylene
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
NuSil Technology’s MED-6640 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Viscosity | 3,000 cP (3,000 mPa*s) |
| Non Volatile Content | 20 % |
| Durometer | 40 Type A |
| Appearance | Translucent |
| Cure | 30 minutes / 25 °C 45 minutes / 75 °C 135 minutes / 150 °C |
| Elongation | 1,000 % |
| Mix Ratio | 1:1 |
| Refractive Index | 1.41 |
| Specific Gravity | 1.41 |
| Stress at Strain | 150 psi / 100 % (1.03 MPa / 100 %) |
| Tear | 300 ppi (52.91 kN/m) |
| Tensile | 1,700 psi (11.72 MPa) |
| Work Time | 24 hours |
| Comment | Ultra High Tear, in Xylene |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.