Description
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
NuSil Technology’s MED-6629 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Cure System | Platinum |
| Viscosity | 400 cP (400 mPa*s) |
| Non Volatile Content | 33 % |
| Durometer | 35 Type A |
| Appearance | Translucent |
| Cure | 75 minutes / 50 °C 60 minutes / 150 °C |
| Elongation | 900 % |
| Mix Ratio | 1:1 |
| Refractive Index | 1.46 |
| Stress at Strain | 305 psi / 300 % (2.1 Mpa / 300 %) |
| Tear | 310 ppi (54.67 kN/m) |
| Tensile | 1,800 psi (12.41 MPa) |
| Comment | High Strength Silicone Dispersion |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.