Implant Line
MED-6629
ADDITION CURE SILICONE DISPERSION
Description
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
NuSil Technology’s MED-6629 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 400 cP (400 mPa*s) |
Non Volatile Content | 33 % |
Durometer | 35 Type A |
Appearance | Translucent |
Cure | 75 minutes / 50 °C 60 minutes / 150 °C |
Elongation | 900 % |
Mix Ratio | 1:1 |
Refractive Index | 1.46 |
Stress at Strain | 305 psi / 300 % (2.1 Mpa / 300 %) |
Tear | 310 ppi (54.67 kN/m) |
Tensile | 1,800 psi (12.41 MPa) |
Comment | High Strength Silicone Dispersion |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.