Description
- Two-part medium viscosity silicone elastomer dispersed in heptane
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
- Useful as a solvent-based adhesive
NuSil™ MED-6615 may be considered for use in human implantation for a period of greater than 29 days
Product Details
| Property | Average Result |
|---|---|
| Cure Type | Heat Cure |
| Mix Ratio | 1:1 |
| Rheology | Flowable / Self Leveling |
| Appearance | Translucent |
| Cure | 30 minutes / RTV |
| Cure System | Platinum |
| Durometer | 72 Type 00 |
| Elongation | 750 % |
| Non Volatile Content | 32 % |
| Stress @ Strain | |
| Substrate | Primerless: Aluminum / Glass / PMMA / Silicone / With Primer: ABS / PEEK / Polycarbonate / Polyimide / Polysulphone / Polyetherimide / Polyurethane / Stainless steel |
| Tear | 120 ppi (21.16 kN/m) |
| Tensile | 1,060 psi (7.31 MPa) |
| Viscosity | 65 cP (65 mPa*s) |
| Comment | Silicone adhesive dispersed in heptane, cures rapidly with heat |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.