Implant Line
MED-6613-6
SILICONE INK (GREEN)
Description
- Strong adhesion to other silicones
- Dispersed in Xylene
- Accelerated cure with introduction of heat
- Cures via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Ideal for use as a marking ink for silicone parts and other components on which the coating must maintain long-term stability
- Use in applications requiring a rapid cure schedule
- Provides high opacity for single print applications
NuSil™ MED-6613-6 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Non Volatile Content | 65 % |
Mix Ratio | 1:1 |
Appearance | Green |
Cure | 5 minutes / 150 °C |
Comment | Heat Curable Ink in Green |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.