Description
- One-part, flowable silicone elastomer dispersion
- Cures at room temperature upon exposure to atmospheric moisture
- Non-corrosive cure system
Applications
- Ideal for use as a marking ink for silicone parts and other components on which the coating must maintain long-term stability
- For use in pad and transfer printing applications
NuSil™ MED-6608-2 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure System | Condensation |
Viscosity | 1,150 cP (1,150 mPa*s) |
Non Volatile Content | 70 % |
Mix Ratio | 1 Part |
Appearance | Black |
Cure | 7 days / RTV |
Specific Gravity | 1.46 |
Tack Free Time | 80 minutes |
Comment | RTV Ink in Black |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.