Description
- A clear silicone gel
- 100% silicone solids (solvent-less)
- A low viscosity liquid that cures to a soft, high tack silicone gel
- 1:1 Mix Ratio (Part A: Part B)
- Supported with applicable biological testing
Applications
- Can be considered for use in wound care applications and scar care products
- Suitable for applications requiring a clear silicone gel with high adhesion
- Ideal for coating various substrates
NuSil Technology’s MED-6360 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Penetration | 14 mm |
Viscosity | 450 cP (450 mPa*s) |
Mix Ratio | 1:1 |
Appearance | Translucent |
Cure | 60 minutes / 140 °C |
Cure System | Platinum |
Tack | 12 psi (0.08 MPa) |
Comment | Very Low Viscosity, Soft Gel |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.