Implant Line
MED-6219
CLEAR, HIGH DUROMETER LOW VISCOSITY SILICONE ELASTOMER
Description
  • Flowable, two-part, clear, high durometer silicone
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
  • Useful in any application requiring a clear, flowable, high durometer material
NuSil Technology’s MED-6219 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer 75 Type A
Viscosity 25,500 cP (25,500 mPa*s)
Work Time 2.5 hours
Tensile 1,300 psi (8.97 MPa)
Appearance Clear
Cure 2 hours / 50 °C
Cure System Platinum
Elongation 65 %
Mix Ratio 1:1
Specific Gravity 1.07
Tear 35 ppi (6.17 kN/m)
Comment Clear, 1.41 R.I., Low Viscosity, High Durometer
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.