Premium Care Line
MED-6033
CLEAR LOW VISCOSITY SILICONE ELASTOMER
Description
  • Two-part, pourable clear, silicone elastomer
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property Average Result
Durometer 50 Type A
Viscosity 72,000 cP (72,000 mPa*s)
Work Time 50 hours
Tensile 730 psi (5.03 MPa)
Appearance Clear
Cure 30 minutes / 150 °C
Cure System Platinum
Elongation 390 %
Mix Ratio 1:1
Refractive Index 1.41
Specific Gravity 1.03
Tear 80 ppi (14.11 kN/m)
Comment 1.41 R.I., Optically Clear, Pourable
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.