Description
- Two-part, pourable, optically clear, silicone elastomer
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
- As an adhesive or encapsulant for applications requiring optical clarity
Product Details
| Property | Average Result |
|---|---|
| Durometer | 40 Type A |
| Viscosity | 65,000 cP (65,000 mPa*s) |
| Work Time | 4 hours |
| Tensile | 640 psi (4.41 MPa) |
| Appearance | Clear |
| Cure | 30 minutes / 150 °C |
| Cure System | Platinum |
| Elongation | 215 % |
| Mix Ratio | 1:1 |
| Refractive Index | 1.43 |
| Tear | 40 ppi (7.05 kN/m) |
| Comment | 1.43 R.I., Optically Clear |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.