MED-6019
OPTICALLY CLEAR, HIGH DUROMETER LOW VISCOSITY SILICONE ELASTOMER
Description
- Flowable, two-part, optically clear, high durometer silicone
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
- Useful in any application requiring a flowable, high durometer material
Product Details
| Property | Average Result |
|---|---|
| Durometer | 75 Type A |
| Viscosity | 25,500 cP (25,500 mPa*s) |
| Work Time | 2.5 hours |
| Tensile | 1,300 psi (8.97 MPa) |
| Appearance | Clear |
| Cure | 2 hours / 50 °C |
| Cure System | Platinum |
| Elongation | 65 % |
| Mix Ratio | 1:1 |
| Specific Gravity | 1.07 |
| Tear | 35 ppi (6.17 kN/m) |
| Comment | Low Viscosity, High Durometer |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.