Premium Care Line
MED-6015
OPTICALLY CLEAR LOW VISCOSITY SILICONE ELASTOMER
Description
  • Two-part, optically clear, solvent free, low viscosity silicone elastomer
  • Cures with heat via addition-cure chemistry
  • 10:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property Average Result
Durometer 50 Type A
Viscosity 3,800 cP (3,800 MPa*s)
Work Time 5.5 hours
Tensile 1,200 psi (8.28 MPa)
Appearance Clear
Cure 15 minutes / 150 °C
Cure System Platinum
Elongation 100 %
Mix Ratio 10:1
Specific Gravity 1.03
Comment 1.41 R.I., Optically Clear, Low Viscosity
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.