Description
- Two-part, translucent silicone system designed for use with injection molding equipment
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a low durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings
- and other precision parts
- Suitable for over-molding applications
- Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
NuSil Technology’s MED-4810 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Durometer | 11 Type A |
| Tensile | 700 psi (4.83 MPa) |
| Elongation | 1075 % |
| Tear | 65 ppi (11.46 kN/m) |
| Appearance | Translucent |
| Cure | 5 minutes / 150 °C |
| Cure System | Platinum |
| Extrusion Rate | 100 g/minute |
| Mix Ratio | 1:1 |
| Specific Gravity | 1.1 |
| Stress at Strain | 45 psi / 200 % (0.31 MPa / 200 %) |
| Work Time | 70 hours |
| Comment | Injection Molding Elastomer |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.