Description
- Two-part, translucent silicone system designed for use with injection molding equipment
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a low durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
- Suitable for over-molding applications
- Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
NuSil Technology’s MED-4805 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
| Property | Average Result |
|---|---|
| Durometer | 7 Type A |
| Tensile | 485 psi (3.34 MPa) |
| Elongation | 1080 % |
| Tear | 70 ppi (12.35 kN/m) |
| Appearance | Translucent |
| Cure | 5 minutes / 150 °C |
| Cure System | Platinum |
| Extrusion Rate | 55 g/minute |
| Mix Ratio | 1:1 |
| Specific Gravity | 1.09 |
| Work Time | 50 hours |
| Comment | Injection Molding Elastomer |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.